Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027937.2 | NP_082213.2 | 18 | 3454 | ||
GenBank mRNA | AK048449.1 | - | 1 | 933 | ||
AK049987.1 | - | 1 | 1271 | |||
AK129327.1 | - | 16 | 3017 | |||
BC059916.1 | - | 9 | 2281 | |||
BC060720.1 | - | 18 | 3218 | |||
BC138443.1 | - | 16 | 3149 | 8861463 | ||
BC145281.1 | - | 16 | 3149 | 9053835 |