Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930503L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172967.2 | NP_766555.1 | 3 | 800 | ||
GenBank mRNA | AK037066.1 | - | 3 | 801 | ||
AK080780.1 | - | 3 | 831 | |||
AK087788.1 | - | 3 | 773 | |||
AK148410.1 | - | 3 | 824 | |||
AK160248.1 | - | 3 | 674 | |||
AK160714.1 | - | 3 | 770 | |||
BC049726.1 | - | 312 | ||||
BC057927.1 | - | 2 | 517 | 5290837 | ||
HQ438967.1 | - | 2 | 312 | |||
HQ438968.1 | - | 2 | 312 | |||
HQ438969.1 | - | 2 | 312 | |||
HQ438970.1 | - | 2 | 312 | |||
HQ438971.1 | - | 2 | 312 | |||
HQ438972.1 | - | 2 | 312 | |||
HQ438973.1 | - | 2 | 321 | |||
HQ438974.1 | - | 2 | 321 | |||
HQ438975.1 | - | 2 | 321 | |||
HQ438976.1 | - | 1 | 271 | |||
HQ438977.1 | - | 1 | 271 | |||
HQ438978.1 | - | 1 | 280 | |||
HQ438979.1 | - | 1 | 271 | |||
HQ438980.1 | - | 1 | 271 | |||
HQ438981.1 | - | 1 | 271 | |||
HQ438982.1 | - | 1 | 271 | |||
HQ438983.1 | - | 1 | 271 | |||
HQ438984.1 | - | 1 | 280 | |||
HQ438985.1 | - | 1 | 271 | |||
HQ438986.1 | - | 1 | 280 | |||
HQ438987.1 | - | 1 | 271 | |||
HQ438988.1 | - | 1 | 271 | |||
HQ438989.1 | - | 1 | 271 | |||
HQ438990.1 | - | 1 | 280 | |||
HQ438991.1 | - | 1 | 280 | |||
HQ438992.1 | - | 1 | 271 | |||
HQ438993.1 | - | 1 | 280 | |||
HQ438994.1 | - | 1 | 271 | |||
HQ438995.1 | - | 1 | 280 | |||
HQ439006.1 | - | 1 | 271 | |||
HQ439007.1 | - | 1 | 280 | |||
HQ439008.1 | - | 1 | 280 | |||
HQ439009.1 | - | 1 | 271 | |||
HQ439010.1 | - | 1 | 280 | |||
HQ439012.1 | - | 1 | 271 |