Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160422.1 | NP_001153894.1 | 4 | 752 | ||
NM_054076.3 | NP_473417.2 | 4 | 752 | |||
GenBank mRNA | AF333980.1 | - | 4 | 751 | ||
AK053318.1 | - | 4 | 535 | |||
AY077682.1 | - | 4 | 744 | |||
BC099435.1 | - | 4 | 585 | 30788813 |