Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001985.3 | NP_001001985.3 | 3 | 2052 | ||
GenBank mRNA | AK147307.1 | - | 3 | 2053 | ||
AK147594.1 | - | 3 | 1331 | |||
AK147703.1 | - | 3 | 1390 | |||
BC034068.1 | - | 1 | 455 | |||
BC075616.1 | - | 3 | 1133 | |||
DQ174094.1 | - | 3 | 1133 |