Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chsy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081163.1 | NP_001074632.1 | 2 | 767 | ||
GenBank mRNA | AK129255.1 | - | 2 | 767 | ||
BC128567.1 | - | 3 | 214 | |||
BC153827.1 | - | 3 | 239 |