Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 39 | 7078 | ||
GenBank mRNA | AK028873.1 | - | 10 | 1081 | ||
AK029836.1 | - | 9 | 945 | |||
BC046526.1 | - | 3 | 194 | |||
BC068298.1 | - | 13 | 1513 | |||
BC096035.1 | - | 2 | 141 |