Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000959.3 | NP_000950.1 | 3 | 1114 | ||
NM_001039585.1 | NP_001034674.1 | 4 | 1185 | |||
GenBank mRNA | AF004021.1 | - | 3 | 999 | ||
AK292845.1 | - | 3 | 1098 | |||
AK312816.1 | - | 3 | 1098 | |||
AY337000.1 | - | 2 | 848 | |||
AY485530.1 | - | 4 | 1156 | |||
BC112965.2 | - | 3 | 1025 | 40025418 | ||
CD013876.1 | - | 4 | 1189 | |||
L24470.1 | - | 3 | 1085 |
Set Membership: |
G-protein coupled receptor |