Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173037.1 | NP_766625.1 | 10 | 1786 | ||
GenBank mRNA | AK047206.1 | - | 5 | 504 | ||
AK054283.1 | - | 10 | 1740 | |||
AK077757.1 | - | 10 | 1771 | |||
AK085030.1 | - | 10 | 1786 | |||
AK173250.1 | - | 10 | 1693 |