Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201406.1 | NP_958808.1 | 3 | 289 | ||
GenBank mRNA | AK033331.1 | - | 3 | 263 | ||
AK043232.1 | - | 3 | 263 | |||
AK045759.1 | - | 3 | 260 | |||
AK053771.1 | - | 3 | 263 | |||
AK142871.1 | - | 3 | 260 | |||
AK151858.1 | - | 3 | 251 | |||
AK152229.1 | - | 3 | 251 | |||
AK153300.1 | - | 3 | 255 | |||
AK158128.1 | - | 3 | 260 | |||
AK166408.1 | - | 3 | 266 | |||
AK169633.1 | - | 3 | 255 | |||
BC058979.1 | - | 3 | 289 | 5716747 |