Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029938.2 | NP_001025109.1 | 5 | 733 | ||
GenBank mRNA | AK139016.1 | - | 5 | 725 | ||
AK160517.1 | - | 3 | 526 | |||
BC038146.1 | - | 1 | 51 | |||
BC051502.1 | - | 2 | 192 | |||
BC116750.1 | - | 5 | 717 | 40126069 | ||
BC119166.1 | - | 5 | 717 | 8733915 |