Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012322.2 | NP_001012322.2 | 10, 11 | 1245 | ||
GenBank mRNA | AK032652.1 | - | 10, 11 | 1245 | ||
AY255623.1 | - | 2, 3 | 134 | |||
BC089355.1 | - | 9, 10 | 1146 | 30551620 |
Set Membership: |
G-protein coupled receptor |