Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012322.2 | NP_001012322.2 | 4, 5 | 626 | ||
GenBank mRNA | AK032652.1 | - | 4, 5 | 628 | ||
BC089355.1 | - | 3, 4 | 527 | 30551620 |
Set Membership: |
G-protein coupled receptor |