Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207013.1 | NP_001193942.1 | 9 | 1170 | ||
NM_001308287.1 | NP_001295216.1 | 7 | 996 | |||
NM_002828.3 | NP_002819.2 | 8 | 1101 | |||
NM_080422.2 | NP_536347.1 | 8 | 1101 | |||
NM_080423.2 | NP_536348.1 | 8 | 1101 | |||
XM_005258124.3 | XP_005258181.1 | 9 | 1168 | |||
XM_005258125.3 | XP_005258182.1 | 9 | 1168 | |||
XM_011525705.2 | XP_011524007.1 | 10 | 1388 | |||
XM_011525706.2 | XP_011524008.1 | 7 | 889 | |||
XM_011525707.2 | XP_011524009.1 | 4 | 496 | |||
XM_017025884.1 | XP_016881373.1 | 8 | 1117 | |||
XM_017025885.1 | XP_016881374.1 | 9 | 1303 | |||
XM_017025886.1 | XP_016881375.1 | 7 | 991 | |||
XM_017025887.1 | XP_016881376.1 | 7 | 942 | |||
XM_017025888.1 | XP_016881377.1 | 7 | 942 | |||
GenBank mRNA | AB209569.1 | - | 7 | 1028 | ||
AK225706.1 | - | 9 | 1049 | |||
AK290649.1 | - | 8 | 983 | |||
AK292570.1 | - | 8 | 1001 | |||
BC008244.1 | - | 8 | 944 | 3872164 | ||
BC016727.1 | - | 8 | 1013 | 4081330 | ||
M25393.1 | - | 8 | 967 |