Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145368.1 | NP_001138840.1 | 8 | 622 | ||
NM_001145369.1 | NP_001138841.1 | 3 | 2671 | |||
NM_001145370.1 | NP_001138842.1 | 3 | 2671 | |||
NM_002829.3 | NP_002820.3 | 8 | 622 | |||
XM_006717197.3 | XP_006717260.1 | 8 | 731 | |||
XM_006717199.3 | XP_006717262.1 | 8 | 756 | |||
XM_006717201.3 | XP_006717264.1 | 7 | 563 | |||
XM_006717202.3 | XP_006717265.1 | 5 | 325 | |||
XM_006717203.3 | XP_006717266.1 | 5 | 284 | |||
XM_006717204.3 | XP_006717267.1 | 8 | 658 | |||
XM_011518888.2 | XP_011517190.1 | 8 | 692 | |||
XM_011518889.2 | XP_011517191.1 | 8 | 1362 | |||
XM_017014955.1 | XP_016870444.1 | 8 | 714 | |||
XM_017014956.1 | XP_016870445.1 | 7 | 570 | |||
XM_017014957.1 | XP_016870446.1 | 8 | 658 | |||
GenBank mRNA | AK295875.1 | - | 7 | 542 | ||
AK304434.1 | - | 8 | 622 | |||
AK316066.1 | - | 8 | 622 | |||
BC126117.1 | - | 7 | 532 | 8991833 | ||
BC143848.1 | - | 7 | 532 | 9052363 | ||
BX648253.1 | - | 3 | 2672 | |||
BX648735.1 | - | 3 | 1106 | |||
DQ104439.1 | - | 7 | 510 | |||
M64572.1 | - | 7 | 533 |