Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301204.1 | NP_001288133.1 | 7 | 941 | ||
NM_001301205.1 | NP_001288134.1 | 6 | 626 | |||
NM_176913.4 | NP_795887.2 | 7 | 1065 | |||
GenBank mRNA | AF488552.1 | - | 7 | 1065 | ||
AK089165.1 | - | 4 | 2679 | |||
AK089244.1 | - | 7 | 927 | |||
AK155099.1 | - | 7 | 893 | |||
AK155308.1 | - | 7 | 892 | |||
AK155417.1 | - | 7 | 896 | |||
AK170094.1 | - | 7 | 893 | |||
AK170970.1 | - | 7 | 893 | |||
AK171044.1 | - | 7 | 893 | |||
AK171122.1 | - | 7 | 893 | |||
AK171155.1 | - | 6 | 606 | |||
AK171196.1 | - | 7 | 893 | |||
AK171268.1 | - | 7 | 893 | |||
AK171503.1 | - | 7 | 893 | |||
AK171606.1 | - | 7 | 909 |