Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199797.1 | NP_001186726.1 | 5 | 693 | ||
NM_002832.3 | NP_002823.3 | 5 | 1539 | |||
NM_080588.2 | NP_542155.1 | 4 | 1032 | |||
NR_037663.1 | 5 | 1486 | ||||
NR_037664.1 | 4 | 1355 | ||||
XM_011509819.1 | XP_011508121.1 | 5 | 874 | |||
XM_011509820.2 | XP_011508122.1 | 5 | 578 | |||
XM_011509821.1 | XP_011508123.1 | 5 | 506 | |||
XM_011509822.2 | XP_011508124.1 | 5 | 377 | |||
XM_011509823.1 | XP_011508125.1 | 5 | 874 | |||
XM_011509824.1 | XP_011508126.1 | 4 | 382 | |||
XM_017001935.1 | XP_016857424.1 | 5 | 871 | |||
XM_017001936.1 | XP_016857425.1 | 5 | 871 | |||
XM_017001937.1 | XP_016857426.1 | 5 | 689 | |||
GenBank mRNA | AF394064.1 | - | 3 | 476 | ||
AK127214.1 | - | 5 | 1532 | |||
AK225920.1 | - | 5 | 537 | |||
AK293186.1 | - | 4 | 382 | |||
AK301051.1 | - | 5 | 507 | |||
AK302869.1 | - | 5 | 687 | |||
BC001746.1 | - | 4 | 1032 | 3543111 | ||
BT009848.1 | - | 4 | 443 | |||
D11327.1 | - | 5 | 547 | |||
M64322.1 | - | 5 | 535 |