Search Thermo Fisher Scientific
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199797.1 | NP_001186726.1 | 3 | 433 | ||
NM_002832.3 | NP_002823.3 | 3 | 1279 | |||
NM_080588.2 | NP_542155.1 | 2 | 772 | |||
XM_011509819.1 | XP_011508121.1 | 3 | 614 | |||
XM_011509820.2 | XP_011508122.1 | 3 | 318 | |||
XM_011509823.1 | XP_011508125.1 | 3 | 614 | |||
XM_017001935.1 | XP_016857424.1 | 3 | 614 | |||
XM_017001936.1 | XP_016857425.1 | 3 | 614 | |||
XM_017001937.1 | XP_016857426.1 | 3 | 432 | |||
GenBank mRNA | AF394064.1 | - | 1 | 216 | ||
AK127214.1 | - | 3 | 1272 | |||
AK225920.1 | - | 3 | 277 | |||
AK301757.1 | - | 3 | 300 | |||
AK301980.1 | - | 3 | 322 | |||
AK302869.1 | - | 3 | 427 | |||
BC001746.1 | - | 2 | 772 | 3543111 | ||
BG340453.1 | - | 3 | 314 | |||
BT009848.1 | - | 2 | 183 | |||
D11327.1 | - | 3 | 287 | |||
DC399028.1 | - | 3 | 437 | |||
M64322.1 | - | 3 | 275 |