Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177318.2 | NP_796292.1 | 5 | 1335 | ||
GenBank mRNA | AK046492.1 | - | 4 | 1382 | ||
AK089104.1 | - | 5 | 1147 | |||
AK157635.1 | - | 4 | 1382 | |||
AY072705.1 | - | 5 | 1345 | |||
BC038037.1 | - | 5 | 1335 | 4190337 |