Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176965.3 | NP_795939.2 | 1 | 107 | ||
GenBank mRNA | DT929470.1 | - | 1 | 107 |