Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Simc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176987.4 | NP_795961.3 | 6 | 3491 | ||
GenBank mRNA | AK028926.1 | - | 6 | 1156 | ||
AK046268.1 | - | 1 | 2865 | |||
AK052561.1 | - | 5 | 1285 | |||
AK166176.1 | - | 5 | 1095 | |||
BC023054.1 | - | 1 | 111 | |||
BC059035.1 | - | 5 | 1750 | |||
BC094550.1 | - | 5 | 1302 |