Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 17, 18 | 1909 | ||
NM_001316677.1 | NP_001303606.1 | 15, 16 | 1582 | |||
NM_001323354.1 | NP_001310283.1 | 15, 16 | 3302 | |||
NM_001323355.1 | NP_001310284.1 | 15, 16 | 1715 | |||
NM_001323356.1 | NP_001310285.1 | 15, 16 | 1709 | |||
NM_001323357.1 | NP_001310286.1 | 12, 13 | 1294 | |||
NM_006504.5 | NP_006495.1 | 16, 17 | 1738 | |||
NM_130435.4 | NP_569119.1 | 13, 14 | 1385 | |||
XM_005252691.2 | XP_005252748.1 | 15, 16 | 1582 | |||
XM_011539994.2 | XP_011538296.1 | 15, 16 | 1697 | |||
XM_011539995.1 | XP_011538297.1 | 15, 16 | 1700 | |||
XM_011539996.1 | XP_011538298.1 | 15, 16 | 1700 | |||
XM_017016467.1 | XP_016871956.1 | 15, 16 | 1700 | |||
XM_017016468.1 | XP_016871957.1 | 15, 16 | 1489 | |||
GenBank mRNA | AJ315969.1 | - | 13, 14 | 1294 | ||
AJ430580.1 | - | 13, 14 | 2143 | |||
AK290414.1 | - | 13, 14 | 1321 | |||
AK291828.1 | - | 15, 16 | 1582 | |||
AK300613.1 | - | 14, 15 | 1475 | |||
BC050062.1 | - | 17, 18 | 1817 | 5274802 | ||
BX648180.1 | - | 10, 11 | 4065 | |||
X54134.1 | - | 16, 17 | 1510 |