Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630001G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177055.3 | NP_796029.2 | 6 | 728 | ||
GenBank mRNA | AK041324.1 | - | 6 | 906 | ||
AK041675.1 | - | 6 | 590 | |||
AK139572.1 | - | 847 | ||||
BC052931.1 | - | 6 | 693 | 1066663 |