Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178778.3 | NP_848893.2 | 9 | 839 | ||
GenBank mRNA | AK044772.1 | - | 9 | 835 | ||
AK082408.1 | - | 10 | 941 | |||
AK083280.1 | - | 9 | 841 | |||
AK163712.1 | - | 9 | 843 | |||
BC132485.1 | - | 9 | 814 | 40130762 | ||
BC144986.1 | - | 10 | 915 | |||
BC144987.1 | - | 9 | 890 | |||
BC144989.1 | - | 10 | 915 |