Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177139.5 | NP_796113.1 | 5 | 628 | ||
GenBank mRNA | AK046251.1 | - | 2 | 184 | ||
AK053620.1 | - | 6 | 759 | |||
BC070462.1 | - | 5 | 649 | 30650880 |