Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172887.2 | NP_766475.2 | 44, 45 | 6783 | ||
GenBank mRNA | AK220506.1 | - | 1, 2 | 405 | ||
CA327618.1 | - | 3, 4 | 770 | |||
CF532608.1 | - | 1, 2 | 486 |