Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113354.1 | NP_001106825.1 | 16 | 2821 | ||
NM_177201.5 | NP_796175.1 | 15 | 2518 | |||
GenBank mRNA | AK036609.1 | - | 15 | 1892 | ||
AK122447.1 | - | 15 | 1912 | |||
AK147939.1 | - | 16 | 2208 | |||
BC138899.1 | - | 15 | 1787 | 8861921 |