Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193644.1 | NP_001180573.1 | 5 | 563 | ||
NM_002857.3 | NP_002848.1 | 5 | 563 | |||
NR_036492.1 | 4 | 453 | ||||
NR_036493.1 | 4 | 477 | ||||
GenBank mRNA | AB018541.1 | - | 5 | 540 | ||
AB062286.1 | - | 4 | 423 | |||
AK300368.1 | - | 4 | 432 | |||
AK303099.1 | - | 4 | 472 | |||
BC000496.2 | - | 5 | 538 | 2820701 | ||
BT006879.1 | - | 5 | 536 | |||
CB993363.1 | - | 580 | ||||
CD557913.1 | - | 5 | 553 | |||
DC405033.1 | - | 4 | 472 | |||
KU178290.1 | - | 5 | 536 | |||
KU178291.1 | - | 4 | 426 | |||
X75535.1 | - | 5 | 546 |