Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5 SIRNA Assays ›
Gene Symbol:
PEX5
Gene Name:
peroxisomal biogenesis factor 5
Gene Aliases:
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
Chromosome Location:
Chr.12: 7188072 - 7218574 on Build GRCh38
UniGene ID:
Hs.567327
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11631
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000319.4
NP_000310.2
13
1448
NM_001131023.1
NP_001124495.1
14
1582
NM_001131024.1
NP_001124496.1
13
1426
NM_001131025.1
NP_001124497.1
14
2003
NM_001131026.1
NP_001124498.1
15
2036
NM_001300789.1
NP_001287718.1
14
1608
XM_005253455.1
XP_005253512.1
13
1492
XM_011520793.2
XP_011519095.1
14
1597
XM_011520795.1
XP_011519097.1
14
2014
XM_011520797.1
XP_011519099.1
14
1507
XM_011520798.1
XP_011519100.1
15
1716
XM_011520799.2
XP_011519101.1
14
2156
XM_011520800.1
XP_011519102.1
14
1602
XM_011520801.2
XP_011519103.1
13
1485
XM_011520802.2
XP_011519104.1
13
1359
XM_017019744.1
XP_016875233.1
13
1941
XM_017019745.1
XP_016875234.1
13
1942
XM_017019746.1
XP_016875235.1
12
1830
XM_017019747.1
XP_016875236.1
12
1809
XM_017019748.1
XP_016875237.1
14
2205
XM_017019749.1
XP_016875238.1
14
1969
XM_017019750.1
XP_016875239.1
14
1483
XM_017019751.1
XP_016875240.1
13
1507
XM_017019752.1
XP_016875241.1
14
1605
XM_017019753.1
XP_016875242.1
13
1491
XM_017019754.1
XP_016875243.1
13
1702
XM_017019755.1
XP_016875244.1
13
2100
XM_017019756.1
XP_016875245.1
13
1372
XM_017019757.1
XP_016875246.1
14
1581
GenBank mRNA
AK225126.1
-
13
1381
AK292256.1
-
15
2036
AK299105.1
-
13
1426
AK299534.1
-
9
937
AK301700.1
-
10
1142
AK302742.1
-
14
1542
AK303515.1
-
14
1608
AK312317.1
-
14
1941
AK316250.1
-
13
1381
BC010621.2
-
14
1471
3872822
KU178292.1
-
13
1432
KU178293.1
-
13
1456
KU178294.1
-
12
1345
U19721.1
-
12
1359
X84899.1
-
13
1474
Z48054.1
-
13
1431
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00165604_m1
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome membrane
negative regulation of protein homotetramerization
peroxisome targeting sequence binding
peroxisome matrix targeting signal-1 binding
protein binding
enzyme binding
small GTPase binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0