Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pkd2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181422.3 | NP_852087.2 | 5 | 1072 | ||
GenBank mRNA | AB290927.1 | - | 5 | 904 | ||
AF271381.1 | - | 5 | 1038 | |||
AK046772.1 | - | 5 | 1073 | |||
BC116297.2 | - | 5 | 1003 | 40094997 | ||
BC116323.2 | - | 5 | 1003 | 40096335 |