Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243738.1 | NP_001230667.1 | 12 | 1807 | ||
NM_004761.4 | NP_004752.1 | 13 | 2004 | |||
XM_011514781.1 | XP_011513083.1 | 10 | 1201 | |||
XM_011514785.2 | XP_011513087.1 | 11 | 1276 | |||
GenBank mRNA | AB012295.1 | - | 12 | 1356 | ||
AK095967.1 | - | 10 | 978 | |||
AK294442.1 | - | 11 | 1460 | |||
AL050259.1 | - | 13 | 1577 | |||
BC000316.1 | - | 7 | 663 | |||
BC005037.1 | - | 10 | 1045 | |||
BC032681.1 | - | 13 | 1559 | 5585507 | ||
BC040989.1 | - | 12 | 1859 | |||
BX648015.1 | - | 12 | 1349 | |||
U68142.1 | - | 7 | 610 |