Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170912.1 | NP_001164383.1 | 18 | 3866 | ||
NM_001170913.1 | NP_001164384.1 | 18 | 3904 | |||
NM_181853.4 | NP_862901.3 | 18 | 3803 | |||
GenBank mRNA | AK147559.1 | - | 18 | 3905 | ||
AK172929.1 | - | 3 | 277 | |||
AY572454.1 | - | 18 | 3866 | |||
AY572455.1 | - | 18 | 3803 | |||
BC138032.1 | - | 17 | 3476 | 8861051 | ||
BC145106.1 | - | 17 | 3476 | 9053654 |