Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ankrd34c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207260.2 | NP_997143.1 | 2 | 1643 | ||
GenBank mRNA | AK045652.1 | - | 2 | 1645 | ||
BC137642.1 | - | 2 | 1313 | 8860661 | ||
BC137643.1 | - | 2 | 1313 | 8860662 |