Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168274.1 | NP_001161746.1 | 1122 | |||
NM_177912.4 | NP_808580.2 | 887 | ||||
GenBank mRNA | AK033603.1 | - | 888 | |||
BC113153.1 | - | 880 | 40058717 | |||
BC113155.1 | - | 1108 | 40058514 |