Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289666.1 | NP_001276595.1 | 10 | 636 | ||
NM_001289667.1 | NP_001276596.1 | 8 | 476 | |||
NM_178801.5 | NP_848916.2 | 10 | 636 | |||
GenBank mRNA | AK028973.1 | - | 186 | |||
AK031517.1 | - | 11 | 722 | |||
AK038419.1 | - | 10 | 614 | |||
AK038472.1 | - | 10 | 581 | |||
AK080479.1 | - | 1 | 2287 | |||
AK144963.1 | - | 10 | 638 | |||
BC094382.1 | - | 10 | 552 | 30605060 | ||
BC137621.1 | - | 10 | 566 | 8860640 | ||
BC144788.1 | - | 8 | 407 |