Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037136.1 | NP_001032213.1 | 14 | 2307 | ||
NM_178119.3 | NP_835220.1 | 15 | 2466 | |||
GenBank mRNA | AK035494.1 | - | 4 | 520 | ||
AK046523.1 | - | 15 | 2427 | |||
AK129289.2 | - | 15 | 2413 | |||
AK147503.1 | - | 15 | 2472 | |||
AK147521.1 | - | 15 | 2958 | |||
AK213954.1 | - | 3 | 300 | |||
BC096588.1 | - | 14 | 2255 |