Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 9 | 1343 | ||
NM_002873.1 | NP_002864.1 | 8 | 1448 | |||
NM_133338.2 | NP_579916.1 | 10 | 1344 | |||
NM_133339.2 | NP_579917.1 | 7 | 901 | |||
NM_133340.2 | NP_579918.1 | 7 | 934 | |||
NM_133341.2 | NP_579919.1 | 7 | 860 | |||
NM_133342.2 | NP_579920.1 | 9 | 1321 | |||
NM_133343.1 | NP_579921.1 | 9 | 1286 | |||
NM_133344.2 | NP_579922.1 | 8 | 1103 | |||
XM_017009679.1 | XP_016865168.1 | 9 | 1163 | |||
XM_017009680.1 | XP_016865169.1 | 8 | 1217 | |||
XM_017009681.1 | XP_016865170.1 | 8 | 1075 | |||
GenBank mRNA | AF017748.1 | - | 8 | 1073 | ||
AF076838.1 | - | 8 | 1448 | |||
AF085736.2 | - | 7 | 901 | |||
AF098533.1 | - | 7 | 880 | |||
AF098534.1 | - | 7 | 806 | |||
AF112263.1 | - | 7 | 925 | |||
AF126424.1 | - | 10 | 1343 | |||
AJ001642.1 | - | 8 | 1112 | |||
AJ004977.1 | - | 8 | 1108 | |||
AK292487.1 | - | 9 | 1321 | |||
AL122068.1 | - | 9 | 1286 | |||
BC032304.1 | - | 7 | 914 | 5170876 | ||
BX537441.1 | - | 9 | 1343 |