Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Intu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175515.5 | NP_780724.3 | 2825 | |||
GenBank mRNA | AK122485.1 | - | 14, 15 | 2365 | ||
BC125523.1 | - | 2772 | 40130038 | |||
BC132387.1 | - | 2772 | 40130664 |