Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 14 | 1928 | ||
NM_001289485.1 | NP_001276414.1 | 15 | 2013 | |||
NM_001289486.1 | NP_001276415.1 | 14 | 1925 | |||
GenBank mRNA | AF338468.1 | - | 12 | 1692 | ||
AF338469.1 | - | 15 | 2007 | |||
AK053319.1 | - | 8 | 1262 | |||
BB538070.2 | - | 4 | 506 | |||
BC050244.1 | - | 12 | 1713 | |||
BC150885.1 | - | 14 | 1890 | 9087796 | ||
BC172032.1 | - | 14 | 1887 | 9054318 |