Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008973.2 | NP_001008973.2 | 7, 8 | 798 | ||
NM_001289485.1 | NP_001276414.1 | 8, 9 | 883 | |||
NM_001289486.1 | NP_001276415.1 | 7, 8 | 795 | |||
GenBank mRNA | AF338468.1 | - | 5, 6 | 562 | ||
AF338469.1 | - | 8, 9 | 877 | |||
BC050244.1 | - | 5, 6 | 582 | |||
BC150885.1 | - | 7, 8 | 760 | 9087796 | ||
BC172032.1 | - | 7, 8 | 757 | 9054318 |