Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029878.1 | NP_001025049.1 | 4 | 386 | ||
GenBank mRNA | BC139127.1 | - | 4 | 437 | 8862149 | |
BC139130.1 | - | 4 | 437 | 8862152 |