Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198658.2 | NP_941060.2 | 11 | 1487 | ||
NM_201619.2 | NP_963913.2 | 11 | 1463 | |||
NM_201622.2 | NP_963916.2 | 11 | 1313 | |||
GenBank mRNA | AY094586.1 | - | 11 | 1487 | ||
AY094587.1 | - | 10 | 1382 | |||
AY094588.1 | - | 10 | 1358 | |||
AY094589.1 | - | 10 | 1339 | |||
AY094590.1 | - | 10 | 1315 | |||
AY094591.1 | - | 10 | 1208 | |||
BC119524.1 | - | 10 | 1382 | 40056073 | ||
BC119525.1 | - | 10 | 1358 | 40056074 |