Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081281.1 | NP_001074750.1 | 2 | 332 | ||
GenBank mRNA | AK052918.1 | - | 2 | 333 |