Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 6430531B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033465.2 | NP_001028637.1 | 4 | 656 | ||
GenBank mRNA | AK131789.1 | - | 4 | 657 | ||
AK164921.1 | - | 4 | 626 | |||
BC025923.1 | - | 3 | 992 | |||
BC145460.1 | - | 4 | 620 | |||
BC145461.1 | - | 4 | 620 | |||
BC145730.1 | - | 4 | 620 | 40131033 | ||
BC171930.1 | - | 4 | 620 |