Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033475.3 | NP_001028647.1 | 5 | 689 | ||
GenBank mRNA | AK147378.1 | - | 5 | 690 | ||
BC150860.1 | - | 5 | 760 | 9087771 |