Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002892.3 | NP_002883.3 | 6 | 554 | ||
NM_023000.2 | NP_075376.2 | 6 | 554 | |||
NM_023001.2 | NP_075377.2 | 6 | 554 | |||
XM_005267964.2 | XP_005268021.1 | 6 | 673 | |||
XM_005267965.2 | XP_005268022.1 | 6 | 673 | |||
XM_017021560.1 | XP_016877049.1 | 6 | 673 | |||
XM_017021561.1 | XP_016877050.1 | 6 | 450 | |||
XM_017021562.1 | XP_016877051.1 | 6 | 673 | |||
XM_017021563.1 | XP_016877052.1 | 6 | 673 | |||
XM_017021564.1 | XP_016877053.1 | 3 | 168 | |||
GenBank mRNA | AK307520.1 | - | 6 | 503 | ||
BC026230.1 | - | 6 | 502 | |||
S66427.1 | - | 6 | 414 |