Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165968.1 | NP_001159440.1 | 6 | 743 | ||
NM_207263.2 | NP_997146.2 | 6 | 740 | |||
GenBank mRNA | AY518699.1 | - | 6 | 694 | ||
BC120840.1 | - | 6 | 735 | 40129763 | ||
BC144902.1 | - | 6 | 743 | 9053450 | ||
BC144927.1 | - | 5 | 624 |