Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102409.1 | NP_001095879.1 | 6 | 899 | ||
NM_001102410.1 | NP_001095880.1 | 6 | 899 | |||
NM_201375.2 | NP_958763.1 | 6 | 899 | |||
GenBank mRNA | AY462056.1 | - | 6 | 815 | ||
AY462057.1 | - | 6 | 815 | |||
AY462059.1 | - | 6 | 815 | |||
AY660572.1 | - | 6 | 815 | |||
BC108335.1 | - | 6 | 866 | 5124483 | ||
BC108934.1 | - | 6 | 757 | |||
BC139720.1 | - | 6 | 807 | 40039854 |