Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201518.4 | NP_958926.1 | 2 | 1620 | ||
GenBank mRNA | AK041311.1 | - | 2 | 1593 | ||
AK076530.1 | - | 1 | 1060 | |||
AK135149.1 | - | 2 | 1621 | |||
AK136973.1 | - | 2 | 1614 | |||
AK140831.1 | - | 2 | 1853 | |||
AK172945.1 | - | 1 | 780 | |||
AY495669.1 | - | 1 | 1059 | |||
BC067058.1 | - | 2 | 1601 | |||
BC096471.1 | - | 2 | 1588 | 3582155 | ||
BC138297.1 | - | 1 | 1068 | 8861316 | ||
BC138298.1 | - | 1 | 1068 | 8861317 |