Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146984.1 | NP_667195.1 | 1 | 160 |
Gene Symbol: Olfr1271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146793.1 | NP_667004.1 | 1 | 160 | ||
GenBank mRNA | BC106779.1 | - | 1 | 160 | 40040295 | |
BC106780.1 | - | 1 | 160 | 40040533 |
Gene Symbol: Olfr1272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146980.1 | NP_667191.1 | 1 | 160 |