Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000322.4 | NP_000313.2 | 1 | 476 | ||
GenBank mRNA | BC074720.2 | - | 1 | 216 | 30915170 | |
M62958.1 | - | 1 | 438 | |||
M73531.1 | - | 1 | 438 |